Merck
  • In vitro and in vivo evaluation of the effect of elevated factor VIII on the thrombogenic process.
In vitro and in vivo evaluation of the effect of elevated factor VIII on the thrombogenic process.
Thrombosis and haemostasis (2012-11-28)
M Golder, J Mewburn, D Lillicrap
ABSTRACT
Factor VIII (FVIII), a procoagulant cofactor, plays a crucial role in the intrinsic coagulation cascade. A causal association between elevated FVIII levels and venous thrombosis incidence has been established; no such association has been confirmed with arterial thrombosis. The independent role of elevated FVIII levels in arteriolar thrombosis was evaluated in a mouse model to determine the thrombogenic potential of elevated levels of FVIII. The in vitro thrombogenic effect of elevated FVIII levels was examined using thrombin-antithrombin (TAT) complex generation and thromboelastography (TEG) assays. The thrombogenic potential of acute and extended elevation of circulating FVIII levels was assessed using ferric chloride induced injury of the cremaster arterioles. The rate of TAT complex formation, and the final concentration of TAT complexes, significantly increased as FVIII levels were elevated from 100% to 400% FVIII activity. TEG analysis of fibrin and clot formation showed that as FVIII levels were elevated, the time to initial fibrin formation decreased and the rate of fibrin formation increased. The acute elevation of circulating FVIII to 400% FVIII activity resulted in significantly decreased times to vessel occlusion. Prolonged elevation of FVIII activity did not significantly affect time to vessel occlusion. In conclusion, acute elevations in FVIII levels result in a non-linear thrombogenic effect, with non-significant increases in thrombogenic risk within the physiological range (FVIII levels up to 200%). Prolonged elevation of plasma FVIII did not further increase the thrombogenic potential of elevated FVIII levels.
Product Description
Millipore
TDA Reagent, suitable for microbiology
Sigma-Aldrich
Iron(III) chloride solution, 0.2 M in 2-methyltetrahydrofuran
Sigma-Aldrich
Iron(III) chloride solution, purum, 45% FeCl3 basis
Sigma-Aldrich
Iron(III) chloride, anhydrous, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Iron(III) chloride hexahydrate, ACS reagent, 97%
Sigma-Aldrich
Iron(III) chloride, sublimed grade, ≥99.9% trace metals basis
Sigma-Aldrich
Iron(III) chloride hexahydrate, reagent grade, ≥98%, chunks
Sigma-Aldrich
Iron(III) chloride hexahydrate, puriss. p.a., ACS reagent, crystallized, 98.0-102% (RT)