Merck
  • Candesartan plus hydrochlorothiazide: an overview of its use and efficacy.
Candesartan plus hydrochlorothiazide: an overview of its use and efficacy.
Expert opinion on pharmacotherapy (2012-11-23)
Amedeo Mugellini, Valentina Nieswandt
ABSTRACT
The 2007 European Society of Cardiology/ European Society of Hypertension guidelines suggests that among the therapeutic possibilities, the use of angiotensin receptor blockers as an important component of antihypertensive combination therapy. Thiazide-type diuretics are usually added in order to increase the therapeutic efficacy in lowering blood pressure (BP). Many authors have demonstrated how effective is a fixed-dose combination of candesartan cilexetil/hydrochlorothiazide in lowering BP, even when compared with other antihypertensive drugs belonging to the same therapeutic category. Moreover, this combination showed good compliance and tolerability and few adverse reactions. The authors analyze the importance of the combination therapy of candesartan cilexetil plus hydrochlorothiazide in the treatment of hypertension. Many authors explain how effective BP control is often achieved only with the combination of drugs with different mechanisms of action, in particular in high risk patients or in those affected with diabetes mellitus or chronic renal disease; in fact, these patients need a more intensive BP control. The fixed-dose combination drug candesartan/hydrochlorothiazide turned out to be a significant alternative in the treatment of hypertension, both for its high efficacy in BP reduction and in prevention of target organ damage, both for its tolerability and patient compliance.
Product Description
Supelco
Hydrochlorothiazide, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hydrochlorothiazide, meets USP testing specifications
USP
Hydrochlorothiazide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hydrochlorothiazide, crystalline
Supelco
Hydrochlorothiazide solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Hydrochlorothiazide for peak identification, European Pharmacopoeia (EP) Reference Standard
Hydrochlorothiazide, European Pharmacopoeia (EP) Reference Standard