Merck
  • Development and validation of a 'universal' HPLC method for pyrethroid quantification in long-lasting insecticidal mosquito nets for malaria control and prevention.
Development and validation of a 'universal' HPLC method for pyrethroid quantification in long-lasting insecticidal mosquito nets for malaria control and prevention.
Tropical medicine & international health : TM & IH (2012-10-31)
David W Jenkins, Arno Hensens, Jacob Lloyd, Michael Payne, Peter Cizmarik, Steve Hamel
ABSTRACT
To outline the development and validation of a universal method for quantifying deltamethrin, permethrin and alpha-cypermethrin levels in a variety of long-lasting insecticidal mosquito nets. Using the HPLC conditions found in the CIPAC method for deltamethrin quantification, the method is based on a simple extraction technique for sample preparation (heating in isooctane at approximately 100 °C for 15 min). The method was validated for linearity, specificity, accuracy, precision, insecticide stability to extraction conditions and required extraction time for insecticide removal. The method was found valid for insecticide quantifications for various types of nets, namely for deltamethrin coated on polyester nets, deltamethrin incorporated into polyethylene nets, permethrin incorporated into polyethylene nets, alpha-cypermethrin coated on polyester nets and alpha-cypermethrin incorporated into polyethylene nets. This method will provide a more simplified approach to testing a variety of nets (different types of fibre) containing deltamethrin, permethrin or alpha-cypermethrin.
Product Description
Supelco
Cypermethrin solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Supelco
α-Cypermethrin, PESTANAL®, analytical standard
Supelco
Cypermethrin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Cypermethrin, PESTANAL®, analytical standard