Merck
  • Site-specific functionalization of RNA molecules by an unnatural base pair transcription system via click chemistry.
Site-specific functionalization of RNA molecules by an unnatural base pair transcription system via click chemistry.
Chemical communications (Cambridge, England) (2012-10-04)
Takumi Ishizuka, Michiko Kimoto, Akira Sato, Ichiro Hirao
ABSTRACT
Toward new biotechnology by genetic alphabet expansion, we developed an efficient site-specific labeling method for large RNA molecules. The combination of unnatural base pair transcription and post-transcriptional modification by click chemistry enables simple RNA labeling with a wide variety of functional groups at desired positions, in a one-pot reaction.
Sigma-Aldrich
Pyrrole-2-carboxaldehyde, 98%
Pricing and availability is not currently available.