Merck
Total synthesis of (-)-teucvidin.
Organic letters (2012-05-19)
Xiaozu Liu, Chi-Sing Lee
ABSTRACT
A concise enantioselective synthesis of (-)-teucvidin has been achieved. Our synthetic strategy involved the diastereoselective Michael/Conia-ene cascade cyclization reaction for rapid establishment of the cis-decalin skeleton with three new stereogenic centers in one pot (72%, single diastereomer), the epoxidation/dealkoxycarbonylation protocol for construction of the fused furanone moiety, and the O-allylation/Claisen rearrangement protocol for construction of the all-carbon quaternary center at C9 of the clerodane skeleton.
Sigma-Aldrich
Decahydronaphthalene, mixture of cis + trans, anhydrous, ≥99%
Sigma-Aldrich
cis-Decahydronaphthalene, 99%
Sigma-Aldrich
trans-Decahydronaphthalene, 99%
Sigma-Aldrich
Decahydronaphthalene, mixture of cis + trans, reagent grade, 98%