Merck
  • Selective synthesis of 3-hydroxy acids from Meldrum's acids using SmI2-H2O.
Selective synthesis of 3-hydroxy acids from Meldrum's acids using SmI2-H2O.
Nature protocols (2012-04-28)
Michal Szostak, Malcolm Spain, David J Procter
ABSTRACT
The single-step synthesis of 3-hydroxy carboxylic acids from readily available Meldrum's acids involves a selective monoreduction using a SmI(2)-H(2)O complex to give products in high crude purity, and it represents a considerable advancement over other methods for the synthesis of 3-hydroxy acids. The protocol includes a detailed guide to the preparation of a single electron-reducing SmI(2)-H(2)O complex and describes two representative examples of the methodology: monoreduction of a fully saturated Meldrum's acid (5-(4-bromobenzyl)-2,2-dimethyl-1,3-dioxane-4,6-dione) and tandem conjugate reduction-selective monoreduction of α,β-unsaturated Meldrum's acid (5-(4-methoxybenzylidene)-2,2-dimethyl-1,3-dioxane-4,6-dione). The protocol for selective monoreduction of Meldrum's acids takes ∼6 h to complete.
Sigma-Aldrich
Chloroform-d, 99.8 atom % D
Pricing and availability is not currently available.
Sigma-Aldrich
1,2-Diiodoethane, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
2,2-Dimethyl-1,3-dioxane-4,6-dione, 98%
Pricing and availability is not currently available.