Merck
  • The management of undesirable cyanobacteria blooms in channel catfish ponds using a constructed wetland: Contribution to the control of off-flavor occurrences.
The management of undesirable cyanobacteria blooms in channel catfish ponds using a constructed wetland: Contribution to the control of off-flavor occurrences.
Water research (2011-10-18)
Fei Zhong, Yunni Gao, Tao Yu, Yongyuan Zhang, Dong Xu, Enrong Xiao, Feng He, Qiaohong Zhou, Zhenbin Wu
ABSTRACT
An exploratory study on the management of undesirable cyanobacteria blooms with respect to off-flavor problems using an integrated vertical-flow constructed wetland (CW) was performed at a small commercial-scale channel catfish farm from 2004 to 2007. The results of the three-year experiment indicated that water treatment by the CW could reduce the possibility of dominance by undesirable cyanobacteria species that often cause off-flavor problems. A detailed investigation in 2007, showed that the concentrations of geosmin, MIB (2-methylisoborneol), and β-cyclocitral in the water of the recirculating pond (4.3ngL(-1), U.D. (undetected) and 0.2ngL(-1), respectively) treated by the CW were significantly lower than those in the control pond (152.6ngL(-1), 63.3ngL(-1) and 254.8ngL(-1), respectively). In addition, the relationships among the cyanobacteria species, the off-flavor compounds and ten environmental variables were explored by canonical correspondence analysis (CCA). The results showed that Oscillatoria sp., Oscillatoria kawamurae and Microcystis aeruginosa were the main sources of off-flavor compounds in the catfish ponds. The successful manipulation of undesirable cyanobacteria species potentially resulted in lower concentrations of odorous compounds in the water of the recirculating pond. An investigation of the concentrations of geosmin and MIB in catfish fillets showed that the levels of odorous compounds were below the OTC (odor threshold concentration) values in the recirculating pond but were above the OTC values from July to October in the control pond. Water recycling by the CW could potentially be one of the best management practices to control off-flavor occurrences in aquaculture.
Sigma-Aldrich
2-Methylisoborneol solution, ~10 mg/mL in methanol, ≥98% (GC)
Sigma-Aldrich
2-Methylisoborneol, ≥98.0% (GC)