Merck
  • Determination of chloropropanols in foods by one-step extraction and derivatization using pressurized liquid extraction and gas chromatography-mass spectrometry.
Determination of chloropropanols in foods by one-step extraction and derivatization using pressurized liquid extraction and gas chromatography-mass spectrometry.
Journal of chromatography. A (2011-08-31)
I Racamonde, P González, R A Lorenzo, A M Carro
ABSTRACT
3-Chloropropane-1,2-diol (3-MCPD) and 1,3-dichloro-2-propanol (1,3-DCP) were determined for the first time in bakery foods using pressurized liquid extraction (PLE) combined with in situ derivatization and GC-MS analysis. This one-step protocol uses N,O-bis(trimethylsilyl)trifluoroacetamide (BSTFA) as silylation reagent. Initially, screening experimental design was applied to evaluate the effects of the variables potentially affecting the extraction process, namely extraction time (min) and temperature (°C), number of cycles, dispersant reagent (diatomaceous earth in powder form and as particulate matter with high pore volume Extrelut NT) and percent of flush ethyl acetate volume (%). To reduce the time of analysis and improve the sensitivity, derivatization of the compounds was performed in the cell extraction. Conditions, such as the volume of BSTFA, temperature and time for the in situ derivatization of analytes using PLE, were optimized by a screening design followed to a Doehlert response surface design. The effect of the in-cell dispersants/adsorbents with diatomaceous earth, Florisil and sodium sulfate anhydrous was investigated using a Box-Behnken design. Using the final best conditions, 1 g of sample dispersed with 0.1 g of sodium sulfate anhydrous and 2.5 g diatomaceous earth was extracted with ethyl acetate. 1 g of Florisil, as clean-up adsorbent, and 70 μL of BSTFA were used for 3 min at 70°C. Under the optimum conditions, the calibration curves showed good linearity (R(2)>0.9994) and precision (relative standard deviation, RSD≤2.4%) within the tested ranges. The limits of quantification for 1,3-DCP and 3-MCDP, 1.6 and 1.7 μg kg(-1), respectively, are far below the established limits in the European and American legislations. The accuracy, precision, linearity, and limits of quantification provided make this analytical method suitable for routine control. The method was applied to the analysis of several toasted bread, snacks, cookies and cereal samples, none of which contained chloropropanols at concentrations above the legislation levels.
Sigma-Aldrich
(S)-(+)-3-Chloro-1,2-propanediol, 97%, optical purity ee: 97% (GLC)
Sigma-Aldrich
1,3-Dichloro-2-propanol, 97%
Sigma-Aldrich
(±)-3-Chloro-1,2-propanediol, 98%
Supelco
(±)-3-Chloro-1,2-propanediol, analytical standard
Sigma-Aldrich
(R)-(−)-3-Chloro-1,2-propanediol, 97%, optical purity ee: 98% (GLC)