Merck
  • Membrane potential depolarization decreases the stiffness of vascular endothelial cells.
Membrane potential depolarization decreases the stiffness of vascular endothelial cells.
Journal of cell science (2011-05-12)
Chiara Callies, Johannes Fels, Ivan Liashkovich, Katrin Kliche, Pia Jeggle, Kristina Kusche-Vihrog, Hans Oberleithner
ABSTRACT
The stiffness of vascular endothelial cells is crucial to mechanically withstand blood flow and, at the same time, to control deformation-dependent nitric oxide release. However, the regulation of mechanical stiffness is not yet understood. There is evidence that a possible regulator is the electrical plasma membrane potential difference. Using a novel technique that combines fluorescence-based membrane potential recordings with atomic force microscopy (AFM)-based stiffness measurements, the present study shows that membrane depolarization is associated with a decrease in the stiffness of endothelial cells. Three different depolarization protocols were applied, all of which led to a similar and significant decrease in cell stiffness, independently of changes in cell volume. Moreover, experiments using the actin-destabilizing agent cytochalasin D indicated that depolarization acts by affecting the cortical actin cytoskeleton. A model is proposed whereby a change of the electrical field across the plasma membrane is directly sensed by the submembranous actin network, regulating the actin polymerization:depolymerization ratio and thus cell stiffness. This depolarization-induced decrease in the stiffness of endothelial cells could play a role in flow-mediated nitric-oxide-dependent vasodilation.
Sigma-Aldrich
Barium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Barium chloride, AnhydroBeads, −10 mesh, 99.95% trace metals basis
Sigma-Aldrich
Barium chloride, 99.999% trace metals basis
Sigma-Aldrich
Barium chloride, 99.9% trace metals basis
Sigma-Aldrich
Barium chloride dihydrate, ≥99.999% trace metals basis
Sigma-Aldrich
Barium chloride dihydrate, ACS reagent, ≥99%
Sigma-Aldrich
Barium chloride dihydrate, ≥99%