Merck
  • Hematopoietic Recovery and Amelioration of Radiation-Induced Lethality by the Vitamin E Isoform δ-Tocotrienol.
Hematopoietic Recovery and Amelioration of Radiation-Induced Lethality by the Vitamin E Isoform δ-Tocotrienol.
Radiation research (2011-03-26)
Merriline M Satyamitra, Shilpa Kulkarni, Sanchita P Ghosh, Conor P Mullaney, Donald Condliffe, Venkataraman Srinivasan
ABSTRACT
δ-Tocotrienol (DT3), a vitamin E isoform, is associated with strong antioxidant and immunomodulatory properties. We confirmed the potent antioxidant activity in membrane systems and showed that DT3 is an effective radiation protector and mitigator. DT3 (4 μM, P < 0.001) inhibited lipid peroxidation in mouse liver microsomes and nitric oxide (NO) formation (20 μM DT3, P < 0.01) in RAW264.7 cells, a murine alveolar macrophage line. In CD2F1 mice exposed to lethal total-body radiation from a (60)Co γ-radiation source, a single subcutaneous (s.c.) injection of DT3 before or after irradiation produced a significant increase in 30-day survival. DT3 was effective from 18.75 to 300 mg/kg (--24 h, P < 0.001). A single dose of 150 or 300 mg/kg DT3 given 24 h before irradiation (radioprotection) resulted in dose reduction factors (DRFs) of 1.19 and 1.27, respectively (P < 0.001). Further, DT3 reduced radiation lethality when administered 2, 6 or 12 h after irradiation, and 150 mg/kg DT3 administered 2 h after exposure conferred a DRF of 1.1 (mitigation). The optimum schedule of 300 mg/kg DT3 24 h prior to 7 Gy significantly reduced pancytopenia compared to irradiated controls (P < 0.05). The large therapeutic potential of and multi-lineage hematopoietic recovery for DT3 warrants further studies.
Product Description
Supelco
δ-Tocotrienol, analytical standard
Sigma-Aldrich
D-δ-Tocotrienol, from rice, ≥65% (GC)