Merck
  • Monamphilectine A, a potent antimalarial β-lactam from marine sponge Hymeniacidon sp: isolation, structure, semisynthesis, and bioactivity.
Monamphilectine A, a potent antimalarial β-lactam from marine sponge Hymeniacidon sp: isolation, structure, semisynthesis, and bioactivity.
Organic letters (2010-10-23)
Edward Avilés, Abimael D Rodríguez
ABSTRACT
Monamphilectine A (1), a new diterpenoid β-lactam alkaloid showing potent antimalarial activity, was isolated in milligram quantities following bioassay-directed extraction of a Puerto Rican marine sponge Hymeniacidon sp. Its structure, established by interpretation of spectral data, was confirmed unequivocally by chemical interconversion and comparison of physical, chemical, and bioactivity data with the natural product. The one-step semisynthesis of monamphilectine A was based on a multicomponent Ugi reaction that also established its absolute stereostructure.
Sigma-Aldrich
Carbenicillin disodium salt, BioReagent, suitable for plant cell culture
Sigma-Aldrich
Carbenicillin disodium, meets USP testing specifications
Sigma-Aldrich
Carbenicillin disodium salt, 89.0-100.5% anhydrous basis