Merck
  • Tuning the shape and thermoelectric property of PbTe nanocrystals by bismuth doping.
Tuning the shape and thermoelectric property of PbTe nanocrystals by bismuth doping.
Nanoscale (2010-07-22)
Qian Zhang, Ting Sun, Feng Cao, Ming Li, Minghui Hong, Jikang Yuan, Qingyu Yan, Huey Hoon Hng, Nianqiang Wu, Xiaogang Liu
ABSTRACT
We report the synthesis of a series of monodispersed Bi-doped PbTe nanocrystals with tunable morphologies by using a doping precursor of bismuth(III) 2-ethylhexanoate. The as-synthesized Pb(1-x)Bi(x)Te (x = 0.005, 0.010, 0.015, 0.020) nanocrystals are characterized by X-ray diffraction, X-ray photoelectron spectroscopy and Hall measurements. The nanocrystals with controlled spherical, cuboctahedral, and cubic shapes were readily prepared by varying the Bi doping concentration. Thermoelectric investigation of these nanocrystals shows that the Bi(3+) doping increases electrical conductivity from 350 to 650 K and changes the Seebeck coefficient sign from positive to negative.
Sigma-Aldrich
2-Ethylhexanoic acid, ≥99%
Sigma-Aldrich
2-Ethylhexanoic acid, ≥99%
Supelco
2-Ethylhexanoic acid, analytical standard
Sigma-Aldrich
2-Ethylhexanoic acid, 99%