Merck
  • Helianthin induces antiproliferative effect on human glioblastoma cells in vitro.
Helianthin induces antiproliferative effect on human glioblastoma cells in vitro.
Journal of neuro-oncology (2010-07-17)
Oana Alexandru, Laura Dragutescu, Ligia Tataranu, Vasile Ciubotaru, Ani Sevastre, Ada Maria Georgescu, Oana Purcaru, Suzana Danoiu, L Magnus Bäcklund, Anica Dricu
ABSTRACT
A major focus of brain cancer research today is to translate understanding of glioma biology into advances in treatment, by exploring the potential of target therapy. Here we investigated the ability of three compounds belonging to the chemical class of azo dyes (methyl red, methyl yellow, and helianthin) to inhibit glioblastoma (GB) cell growth in vitro. Our results showed that helianthin induced cytotoxicity in two GB cell cultures, cell lines 18 and 38, whereas methyl red and methyl yellow were not cytotoxic. The effect of helianthin on EGFR, IGF-1R, and their common intracellular signaling via PI3-K and ERK1/2 was also analyzed. Treatment with helianthin down-regulated EGFR and IGF-1R activity in both cell lines. Helianthin treatment blocked ERK1/2 phosphorylation without affecting PI3K activity in cell line 18 and reduced both PI3K and ERK1/2 in GB 38 cell line. The cell death was accompanied by degradation of PARP without affecting BCL2 expression in both GB cell cultures. Because of the genetic heterogeneity of malignant gliomas, we tested the effect of helianthin on other two primary GB lines (11 and 15) and two early-passage GB cultures (BT1GB and BT2GB), to assess the general nature of the anti-tumor effect of the drug in GB cells. We found that helianthin treatment induced cell death in all the GB cell cultures analyzed. To our knowledge, this is the first report indicating that helianthin can reduce GB cell growth.
Sigma-Aldrich
Methyl Red sodium salt, Crystalline
Sigma-Aldrich
Methyl Red sodium salt, ACS reagent, Dye content 95 %
Sigma-Aldrich
Methyl Red, ACS reagent, crystalline
Millipore
Methyl Red solution, suitable for microbiology