Merck
  • Oligopyrenotides: abiotic, polyanionic oligomers with nucleic acid-like structural properties.
Oligopyrenotides: abiotic, polyanionic oligomers with nucleic acid-like structural properties.
Journal of the American Chemical Society (2010-05-13)
Robert Häner, Florian Garo, Daniel Wenger, Vladimir L Malinovskii
ABSTRACT
Oligopyrenotides, abiotic oligomers that exhibit significant structural analogies to the nucleic acids, are described. They are composed of achiral, phosphodiester-linked pyrene building blocks and a single chiral 1,2-diaminocyclohexane unit. These oligomers form stable hybrids in aqueous solution. Hybridization is based on stacking interactions of the pyrene building blocks. They show thermal denaturation/renaturation behavior that closely resembles DNA and RNA hybridization. In addition, oligopyrenotides display salt-concentration-dependent structural polymorphism. Thus, they possess a number of structural attributes that are typical of nucleic acids and therefore may serve as model systems for the design of artificial self-replicating systems.
Sigma-Aldrich
(±)-trans-1,2-Diaminocyclohexane, 99%
Sigma-Aldrich
(1R,2R)-(−)-1,2-Diaminocyclohexane, 98%
Sigma-Aldrich
1,2-Diaminocyclohexane, mixture of cis and trans, 99%
Sigma-Aldrich
(1S,2S)-(+)-1,2-Diaminocyclohexane, 98%