Merck
  • Solvent induced reactivity of 3,5-dimethylpyrazole towards zinc (II) carboxylates.
Solvent induced reactivity of 3,5-dimethylpyrazole towards zinc (II) carboxylates.
Dalton transactions (Cambridge, England : 2003) (2009-09-04)
Rupam Sarma, Dipjyoti Kalita, Jubaraj B Baruah
ABSTRACT
The reactions of 3,5-dimethylpyrazole with zinc(II)acetate dihydrate and varieties of aromatic carboxylic acids led to formation of mono-nuclear zinc complexes of composition [Zn(HDMP)2(RCO2)2] (R = C6H5, p-CH3-C6H4, p-NO2-C6H4 etc. HDMP = 3,5-dimethylpyrazole) in methanol, whereas the same reactants in dimethylformamide (DMF) gave binuclear 3,5-dimethylpyrazolato bridged zinc carboxylate complexes containing monodentate 3,5-dimethylpyraozole ligands with composition [Zn2(mu-DMP)2(HDMP)2(RCO2)2]. The mononuclear complexes can be converted to the corresponding binuclear complexes by simply dissolving in DMF. The reaction of zinc(II)acetate dihydrate with p-nitrobenzoic acid and 3,5-dimethylpyrazole in different solvents gave solvated mononuclear complexes of the corresponding solvent. All these solvated complexes having the core [Zn(HDMP)2(p-NO2-C6H4CO2)2] contain two structurally independent molecules in the asymmetric unit (Z' = 2).
Sigma-Aldrich
3,5-Dimethylpyrazole, produced by Wacker Chemie AG, Burghausen, Germany, Wacker Chemie AG, ≥99.0% (GC)
Sigma-Aldrich
3,5-Dimethylpyrazole, 99%