Merck
  • Ab initio study of the barriers to methyl torsion and torsional frequencies of acetyl molecules.
Ab initio study of the barriers to methyl torsion and torsional frequencies of acetyl molecules.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2005-04-12)
Stephen Bell
ABSTRACT
A wide range of ab initio and hybrid density functional methods and basis sets have been employed to calculate the barriers to methyl internal rotation in a range of molecules with the acetyl moiety. Comparison is made of the computed torsional frequency with the experimental torsional frequency, nu(obs), for each molecule. With the MP2/6-311+G(3df,2p) combination of method and basis set, the agreement is better than 4 cm-1 for most of the molecules, where nu(obs) or the V3 barrier is well-determined experimentally.
Sigma-Aldrich
Acetyl chloride, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Acetyl chloride, reagent grade, 98%
Sigma-Aldrich
Acetyl chloride, reagent grade, 98%
Sigma-Aldrich
Acetyl bromide, 99%
Sigma-Aldrich
Acetyl isothiocyanate, ≥97.0% (GC)