Merck
  • Stereochemical features of Lewis acid-promoted glycosidations involving 4'-spiroannulated DNA building blocks.
Stereochemical features of Lewis acid-promoted glycosidations involving 4'-spiroannulated DNA building blocks.
The Journal of organic chemistry (2004-10-23)
Leo A Paquette, Christopher K Seekamp, Alexandra L Kahane, David G Hilmey, Judith Gallucci
ABSTRACT
Tin tetrachloride-catalyzed glycosidation of persilylated nucleobases with acetate donor 6 in CH(2)Cl(2) solution followed by deprotection gave rise very predominantly to alpha-spironucleosides. These stereochemical assignments stem from the determination of NOE interactions and an X-ray crystallographic analysis of the latter product. Computational studies revealed that these results are consistent with the fact that the C5' substituent shields the beta-face of the oxonium ion involved in the coupling reaction while the C3' substituent is projected away from the alpha-underside. Attack from the more open direction is therefore kinetically favored. Entirely comparable calculations suggested that donor 19 should behave comparably. Experimentation involving this donor gave results consistent with this model although more equitable alpha/beta spironucleoside product ratios were seen when acetonitrile was employed as the reaction medium.
Sigma-Aldrich
Tin(IV) chloride, 98%
Sigma-Aldrich
Tin(IV) chloride solution, 1.0 M in methylene chloride
Sigma-Aldrich
Tin(IV) chloride, 99.995% trace metals basis