Merck
Optical rotation of noncovalent aggregates.
Journal of the American Chemical Society (2003-12-18)
Michael-Rock Goldsmith, Nilu Jayasuriya, David N Beratan, Peter Wipf
ABSTRACT
Dilute solutions of (R)-(-)-pantolactone in CCl4 were studied by polarimetry in conjunction with theoretical calculations of [alpha]D. Our data demonstrate that the self-association of a chiral solute results in a change in [alpha]D that can be accounted for by the presence of hydrogen-bonded dimeric species. The theoretical analysis predicts a concentration-dependent specific rotation in good agreement with experiment. Further exploration of monomer and dimer [alpha]D differences, through atomic map analysis, reveals large contributions to [alpha]D from the hydrogen-bonded hydroxyl groups in the tightly-coupled dimer. This study extends the computation of chiroptical properties to the accurate concentration-dependent prediction of [alpha]D for noncovalently interacting self-associating species.
Sigma-Aldrich
(S)-(+)-Pantolactone, 97%
Sigma-Aldrich
DL-α-Hydroxy-β,β-dimethyl-γ-butyrolactone, purum, ≥97.0% (T)
Sigma-Aldrich
D-(−)-Pantolactone, 99%