Merck
  • Bioprospecting for novel hydroxyoxylipins in fungi: presence of 3-hydroxy palmitic acid in Saccharomycopsis malanga.
Bioprospecting for novel hydroxyoxylipins in fungi: presence of 3-hydroxy palmitic acid in Saccharomycopsis malanga.
Antonie van Leeuwenhoek (2002-02-06)
O Sebolai, J L Kock, C H Pohl, D P Smith, P J Botes, E E Pretorius, P W Van Wyk, S Nigam
ABSTRACT
Electron microscopy studies indicated that the major oxylipin 3-hydroxy palmitic acid (16:0) was associated with aggregating vegetative cells and formed a web-like structure around these cells. Cross sections through this structure showed a hydrophilic outer layer and a more hydrophobic inner layer suggesting that the web-like structure is in fact tube-like micelles. This information sheds more light on the role of these hydroxyoxylipins in fungi.
Sigma-Aldrich
DL-β-Hydroxypalmitic acid, ≥98%