Merck
  • RNF220 is an E3 ubiquitin ligase for AMPA receptors to regulate synaptic transmission.
RNF220 is an E3 ubiquitin ligase for AMPA receptors to regulate synaptic transmission.
Science advances (2022-10-01)
Pengcheng Ma, Li Pear Wan, Yuwei Li, Chun-Hui He, Ning-Ning Song, Shiping Zhao, Huishan Wang, Yu-Qiang Ding, Bingyu Mao, Nengyin Sheng
ABSTRACT
The accurate expression of postsynaptic AMPA receptors (AMPARs) is critical for information processing in the brain, and ubiquitination is a key regulator for this biological process. However, the roles of E3 ubiquitin ligases in the regulation of AMPARs are poorly understood. Here, we find that RNF220 directly interacts with AMPARs to meditate their polyubiquitination, and RNF220 knockout specifically increases AMPAR protein levels, thereby enhancing basal synaptic activity while impairing synaptic plasticity. Moreover, depending on its E3 ubiquitin ligase activity, RNF220 represses AMPAR-mediated excitatory synaptic responses and their neuronal surface expression. Furthermore, learning and memory are altered in forebrain RNF220-deficient mice. In addition, two neuropathology-related RNF220 variants fail to repress excitatory synaptic activity because of the incapability to regulate AMPAR ubiquitination due to their attenuated interaction. Together, we identify RNF220 as an E3 ubiquitin ligase for AMPARs and establish its substantial role in excitatory synaptic transmission and brain function.
Sigma-Aldrich
Anti-HA antibody, Mouse monoclonal, clone HA-7, purified from hybridoma cell culture
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-RNF220 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, Type II, lyophilized powder, Protein ≥80 %, ≥2,000 units/mg protein
Sigma-Aldrich
Laminin from Engelbreth-Holm-Swarm murine sarcoma basement membrane, 1-2 mg/mL in Tris-buffered saline, 0.2 μm filtered, BioReagent, suitable for cell culture