Merck
  • p53 is a central regulator driving neurodegeneration caused by C9orf72 poly(PR).
p53 is a central regulator driving neurodegeneration caused by C9orf72 poly(PR).
Cell (2021-01-23)
Maya Maor-Nof, Zohar Shipony, Rodrigo Lopez-Gonzalez, Lisa Nakayama, Yong-Jie Zhang, Julien Couthouis, Jacob A Blum, Patricia A Castruita, Gabriel R Linares, Kai Ruan, Gokul Ramaswami, David J Simon, Aviv Nof, Manuel Santana, Kyuho Han, Nasa Sinnott-Armstrong, Michael C Bassik, Daniel H Geschwind, Marc Tessier-Lavigne, Laura D Attardi, Thomas E Lloyd, Justin K Ichida, Fen-Biao Gao, William J Greenleaf, Jennifer S Yokoyama, Leonard Petrucelli, Aaron D Gitler
ABSTRACT
The most common genetic cause of amyotrophic lateral sclerosis (ALS) and frontotemporal dementia (FTD) is a GGGGCC repeat expansion in the C9orf72 gene. We developed a platform to interrogate the chromatin accessibility landscape and transcriptional program within neurons during degeneration. We provide evidence that neurons expressing the dipeptide repeat protein poly(proline-arginine), translated from the C9orf72 repeat expansion, activate a highly specific transcriptional program, exemplified by a single transcription factor, p53. Ablating p53 in mice completely rescued neurons from degeneration and markedly increased survival in a C9orf72 mouse model. p53 reduction also rescued axonal degeneration caused by poly(glycine-arginine), increased survival of C9orf72 ALS/FTD-patient-induced pluripotent stem cell (iPSC)-derived motor neurons, and mitigated neurodegeneration in a C9orf72 fly model. We show that p53 activates a downstream transcriptional program, including Puma, which drives neurodegeneration. These data demonstrate a neurodegenerative mechanism dynamically regulated through transcription-factor-binding events and provide a framework to apply chromatin accessibility and transcription program profiles to neurodegeneration.
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®
Sigma-Aldrich
Anti-GAPDH antibody, Mouse monoclonal, clone GAPDH-71.1, purified from hybridoma cell culture
Sigma-Aldrich
Anti-C9ORF72/C9RANT (Poly-GR), serum, from rabbit
Sigma-Aldrich
Cycloheximide, ≥95% (HPLC)
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-TDP-43 (C-terminal) antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse