Merck
  • Identification and analysis of OsttaDSP, a phosphoglucan phosphatase from Ostreococcus tauri.
Identification and analysis of OsttaDSP, a phosphoglucan phosphatase from Ostreococcus tauri.
PloS one (2018-01-24)
Julieta B Carrillo, Diego F Gomez-Casati, Mariana Martín, Maria V Busi
ABSTRACT
Ostreococcus tauri, the smallest free-living (non-symbiotic) eukaryote yet described, is a unicellular green alga of the Prasinophyceae family. It has a very simple cellular organization and presents a unique starch granule and chloroplast. However, its starch metabolism exhibits a complexity comparable to higher plants, with multiple enzyme forms for each metabolic reaction. Glucan phosphatases, a family of enzymes functionally conserved in animals and plants, are essential for normal starch or glycogen degradation in plants and mammals, respectively. Despite the importance of O. tauri microalgae in evolution, there is no information available concerning the enzymes involved in reversible phosphorylation of starch. Here, we report the molecular cloning and heterologous expression of the gene coding for a dual specific phosphatase from O. tauri (OsttaDSP), homologous to Arabidopsis thaliana LSF2. The recombinant enzyme was purified to electrophoretic homogeneity to characterize its oligomeric and kinetic properties accurately. OsttaDSP is a homodimer of 54.5 kDa that binds and dephosphorylates amylopectin. Also, we also determined that residue C162 is involved in catalysis and possibly also in structural stability of the enzyme. Our results could contribute to better understand the role of glucan phosphatases in the metabolism of starch in green algae.
Sigma-Aldrich
DL-Cysteine, technical grade
Millipore
Sea salts, NutriSelect® Basic
Sigma-Aldrich
p-Nitrophenyl Phosphate Liquid Substrate System, liquid
Sigma-Aldrich
4-Nitrophenyl phosphate di(tris) salt, phosphatase substrate