Merck
  • Destabilase-lysozyme of medicinal leech. Multifunctionality of recombinant protein.
Destabilase-lysozyme of medicinal leech. Multifunctionality of recombinant protein.
Biochemistry. Biokhimiia (2010-11-17)
L L Zavalova, V N Lazarev, S A Levitsky, T G Yudina, I P Baskova
ABSTRACT
Preparation and purification of a recombinant protein are described along with characteristics of its specific (for ε-(γ-Glu)-Lys and D-dimer substrates) and nonspecific (for L-γ-Glu-pNA) isopeptidase activities; the absence of peptidase function for α-(α-Glu)-Lys substrate is noted. It is shown that the protein exhibits muramidase (cell walls of Micrococcus lysodeikticus) and specific glycosidase activities. The latter was determined towards the fluorogenic substrate 4-methylumbelliferyl-tetra-N-acetyl-β-chitotetraoxide. Antimicrobial activity of recombinant destabilase-lysozyme protein (recDest-Lys) and its 11-membered amphipathic peptide was revealed towards cells of the strict anaerobic Archaean Methanosarcina barkeri, whose cell walls contain no murein. Possible mechanisms of the effect of recDest-Lys on these cells are discussed.
Sigma-Aldrich
γ-Glu-ε-Lys