Merck
  • The Role of Matrix Metalloproteinases and Tissue Inhibitors of Metalloproteinases in Duchenne Muscular Dystrophy Cardiomyopathy.
The Role of Matrix Metalloproteinases and Tissue Inhibitors of Metalloproteinases in Duchenne Muscular Dystrophy Cardiomyopathy.
Journal of cardiac failure (2019-02-15)
Jonathan H Soslow, Meng Xu, James C Slaughter, Kimberly Crum, Joshua D Chew, W Bryan Burnette, Yan Ru Su, Kelsey Tomasek, David A Parra, Larry W Markham
ABSTRACT
Cardiomyopathy is the leading cause of death in Duchenne muscular dystrophy (DMD). Standard cardiac biomarkers are poor indicators of DMD cardiovascular disease. Matrix metalloproteinases (MMPs) and tissue inhibitors of metalloproteinases (TIMPs) regulate collagen turnover. Given the cardiac fibrosis seen in DMD, we hypothesized that MMPs and TIMPs correlate with severity of DMD cardiomyopathy. Prospectively enrolled DMD subjects (n = 42) underwent cardiac magnetic resonance imaging for function and late gadolinium enhancement (LGE), including LGE severity from 0 (no LGE) to 4 (severe). Serum from DMD and healthy male control subjects (n = 15) analyzed for MMPs 1, 2, 3, 7, 9, and 10 and TIMPs 1-4. MMP1, MMP7, and MMP10 were higher in DMD than in control (respectively, median 5080 pg/mL vs 2120 pg/mL [P = .007], 2170 pg/mL vs 1420 pg/mL [P < .001], and 216 pg/mL vs 140pg/mL [P = .040]); TIMP4 was lower in DMD (124 pg/mL vs 263 pg/mL; P = .046). Within DMD, MMP7 correlated inversely with left ventricular ejection fraction (r = -0.40; P = .012) and directly with strain (r = 0.54; P = .001) and LGE severity (r = 0.47; P = .003). MMP7 was higher in DMD patients with LGE compared with those without LGE and control subjects (P < .001). Multiple MMPs are elevated in DMD compared with control subjects. MMP7 is related to DMD cardiac dysfunction and myocardial fibrosis, possibly through remodeling of the extracellular matrix.
Product Description
Millipore
MILLIPLEX® Human MMP Magnetic Bead Panel 2 - Immunology Multiplex Assay, Matrix Metalloproteinase Bead-Based Multiplex Assays using the Luminex technology enable the simultaneous analysis of multiple MMPs biomarkers in human serum, plasma and cell culture samples.
Millipore
MILLIPLEX® Human MMP Magnetic Bead Panel 1 - Immunology Multiplex Assay, Matrix Metalloproteinase Bead-Based Multiplex Assays using the Luminex technology enable the simultaneous analysis of multiple MMPs biomarkers in human serum, plasma and cell culture samples.
Millipore
MILLIPLEX® Human TIMP Magnetic Bead Panel 2 - Immunology Multiplex Assay, TIMP Bead-Based Multiplex Assays using the Luminex technology enables the simultaneous analysis of multiple TIMP biomarkers in human cell/tissue culture supernatant samples.