Merck
  • Control of multiciliogenesis by miR-34/449 in the male reproductive tract through enforcing cell cycle exit.
Control of multiciliogenesis by miR-34/449 in the male reproductive tract through enforcing cell cycle exit.
Journal of cell science (2021-05-12)
Yu-Jie Wu, Yue Liu, Yan-Qin Hu, Li Wang, Fu-Rong Bai, Chen Xu, Jing-Wen Wu
ABSTRACT
Multiciliated cells (MCCs) are terminally differentiated postmitotic cells that possess hundreds of motile cilia on their apical surface. Defects in cilia formation are associated with ciliopathies that affect many organs. In this study, we tested the role and mechanism of the miR-34/449 family in the regulation of multiciliogenesis in EDs using an miR-34b/c-/-; miR-449-/- double knockout (dKO) mouse model. MiR-34b/c and miR-449 depletion led to a reduced number of MCCs and abnormal cilia structure in the EDs starting from postnatal day (P)14. However, abnormal MCC differentiation in the dKO EDs could be observed as early as P7. RNA-seq analyses revealed that the aberrant development of MCCs in the EDs of dKO mice was associated with the upregulation of genes involved in cell cycle control. Using a cyclin-dependent kinase inhibitor to force cell cycle exit promoted MCC differentiation, and partially rescued the defective multiciliogenesis in the EDs of dKO mice. Taken together, our results suggest that miR-34b/c and miR-449 play an essential role in multiciliogenesis in EDs by regulating cell cycle exit.
Sigma-Aldrich
Monoclonal Anti-Tubulin, Acetylated antibody produced in mouse, clone 6-11B-1, ascites fluid
Sigma-Aldrich
Anti-Cyclin B1 antibody produced in rabbit, affinity isolated antibody