Merck
Developmental cell programs are co-opted in inflammatory skin disease.
Science (New York, N.Y.) (2021-01-23)
Gary Reynolds, Peter Vegh, James Fletcher, Elizabeth F M Poyner, Emily Stephenson, Issac Goh, Rachel A Botting, Ni Huang, Bayanne Olabi, Anna Dubois, David Dixon, Kile Green, Daniel Maunder, Justin Engelbert, Mirjana Efremova, Krzysztof Polański, Laura Jardine, Claire Jones, Thomas Ness, Dave Horsfall, Jim McGrath, Christopher Carey, Dorin-Mirel Popescu, Simone Webb, Xiao-Nong Wang, Ben Sayer, Jong-Eun Park, Victor A Negri, Daria Belokhvostova, Magnus D Lynch, David McDonald, Andrew Filby, Tzachi Hagai, Kerstin B Meyer, Akhtar Husain, Jonathan Coxhead, Roser Vento-Tormo, Sam Behjati, Steven Lisgo, Alexandra-Chloé Villani, Jaume Bacardit, Philip H Jones, Edel A O'Toole, Graham S Ogg, Neil Rajan, Nick J Reynolds, Sarah A Teichmann, Fiona M Watt, Muzlifah Haniffa
ABSTRACT
The skin confers biophysical and immunological protection through a complex cellular network established early in embryonic development. We profiled the transcriptomes of more than 500,000 single cells from developing human fetal skin, healthy adult skin, and adult skin with atopic dermatitis and psoriasis. We leveraged these datasets to compare cell states across development, homeostasis, and disease. Our analysis revealed an enrichment of innate immune cells in skin during the first trimester and clonal expansion of disease-associated lymphocytes in atopic dermatitis and psoriasis. We uncovered and validated in situ a reemergence of prenatal vascular endothelial cell and macrophage cellular programs in atopic dermatitis and psoriasis lesional skin. These data illustrate the dynamism of cutaneous immunity and provide opportunities for targeting pathological developmental programs in inflammatory skin diseases.
Sigma-Aldrich
L-Glutamine solution, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Dulbecco′s Phosphate Buffered Saline, Modified, without calcium chloride and magnesium chloride, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Penicillin-Streptomycin, with 10,000 units penicillin and 10 mg streptomycin per mL in 0.9% NaCl, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, for molecular biology, 0.5 M in H2O, DNase, RNase, NICKase and protease, none detected
Sigma-Aldrich
Triton X-100, for molecular biology
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Triton X-100, laboratory grade