Merck
  • Experimental design of a culture approach for corneal endothelial cells of New Zealand white rabbit.
Experimental design of a culture approach for corneal endothelial cells of New Zealand white rabbit.
Heliyon (2020-10-20)
María Dolores Montalvo-Parra, Isaac Alejandro Vidal-Paredes, Cesar E Calzada-Rodríguez, Italia Tatnaí Cárdenas-Rodríguez, Guiomar Farid Torres-Guerrero, Daniela Gómez-Elizondo, Mariana López-Martínez, Judith Zavala, Jorge E Valdez-García
ABSTRACT
The harvesting of corneal endothelial cells (CEC) has received special attention due to its potential as a therapy for corneal blindness. The main challenges are related to the culture media formulation, cellular density at the primary isolation, and the number of passages in which CEC can retain their functional characteristics. To alternate different media formulations to harvest CEC has an impact on the cellular yield and morphology. Therefore, we analyzed four different sequences of growth factor-supplemented Stimulatory (S) and non-supplemented Quiescent (Q) media, upon passages to find the optimal S-Q culture sequence. We assessed cell yield, morphology, procollagen I production, Na+/K+-ATPase function, and the expression of ZO-1 and Na+/K+-ATPase. Our results show SQSQ and SQQQ sequences with a balance between an improved cell yield and hexagonal morphology rate. CEC cultured in the SQQQ sequence produced procollagen I, showed Na+/K+-ATPase function, and expression of ZO-1 and Na+/K+-ATPase. Our study sets a culture approach to guarantee CEC expansion, as well as functionality for their potential use in tissue engineering and in vivo analyses. Thus, the alternation of S and Q media improves CEC culture. SQQQ sequence demonstrated CEC proliferation and lower the cost implied in SQSQ sequences. We discarded the use of pituitary extract and ROCK inhibitors as essential for CEC proliferation.
Sigma-Aldrich
EGF Protein, Human Recombinant Animal Free, This Recombinant animal free human EGF is manufactured using all non-animal reagents.
Sigma-Aldrich
Poly-D-lysine hydrobromide, average mol wt 30,000-70,000, lyophilized powder, γ-irradiated, BioReagent, suitable for cell culture
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
Collagenase from Clostridium histolyticum, for general use, Type I, ≥125 CDU/mg solid
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, fatty acid free, essentially globulin free, pH 7, ≥98%