Merck
  • The immunodominant Brugia malayi paramyosin as a marker of current infection with Wuchereria bancrofti adult worms.
The immunodominant Brugia malayi paramyosin as a marker of current infection with Wuchereria bancrofti adult worms.
Infection and immunity (1998-05-29)
S Langy, C Plichart, P Luquiaud, S A Williams, L Nicolas
ABSTRACT
The full-length cDNA sequence encoding Brugia malayi L3 paramyosin has been isolated by immunoscreening a cDNA library with a mouse antiserum raised against Wuchereria bancrofti L3 infective larvae. A recombinant truncated form of paramyosin was expressed as a glutathione S-transferase fusion protein and used to evaluate humoral responses of adults from a W. bancrofti-endemic area in French Polynesia according to their parasitological status. Immunoglobulin G4 (IgG4) preferentially bound to paramyosin in W. bancrofti-parasitized individuals, in contrast to unparasitized individuals, who harbored neither microfilaria nor Og4C3 adult worm circulating antigen. Reduction of the anti-paramyosin IgG4 titer following combined chemotherapy with diethylcarbamazine and ivermectin was significantly correlated with a reduction in the adult worm burden. This indicates that the presence of paramyosin-reactive IgG4 is associated with the presence of parasites and that reduction can be used as an immunological marker for W. bancrofti clearance.
Sigma-Aldrich
Monoclonal Anti-Human IgG (Fc specific) antibody produced in mouse, clone HP-6017, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Human IgG4 (pFc′ specific) antibody produced in mouse, clone HP-6023, ascites fluid
Sigma-Aldrich
Mouse Anti-Human IgG2 Antibody, clone HP6002, Fc, clone HP6002, Chemicon®, from mouse
Sigma-Aldrich
Monoclonal Anti-Human IgG1 (Fc specific) antibody produced in mouse, clone HP-6001, ascites fluid