Merck
Targeting the Apoa1 locus for liver-directed gene therapy.
Molecular therapy. Methods & clinical development (2021-06-19)
Marco De Giorgi, Ang Li, Ayrea Hurley, Mercedes Barzi, Alexandria M Doerfler, Nikitha A Cherayil, Harrison E Smith, Jonathan D Brown, Charles Y Lin, Karl-Dimiter Bissig, Gang Bao, William R Lagor
ABSTRACT
Clinical application of somatic genome editing requires therapeutics that are generalizable to a broad range of patients. Targeted insertion of promoterless transgenes can ensure that edits are permanent and broadly applicable while minimizing risks of off-target integration. In the liver, the Albumin (Alb) locus is currently the only well-characterized site for promoterless transgene insertion. Here, we target the Apoa1 locus with adeno-associated viral (AAV) delivery of CRISPR-Cas9 and achieve rates of 6% to 16% of targeted hepatocytes, with no evidence of toxicity. We further show that the endogenous Apoa1 promoter can drive robust and sustained expression of therapeutic proteins, such as apolipoprotein E (APOE), dramatically reducing plasma lipids in a model of hypercholesterolemia. Finally, we demonstrate that Apoa1-targeted fumarylacetoacetate hydrolase (FAH) can correct and rescue the severe metabolic liver disease hereditary tyrosinemia type I. In summary, we identify and validate Apoa1 as a novel integration site that supports durable transgene expression in the liver for gene therapy applications.
Sigma-Aldrich
Anti-FAH antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
ApopTag Peroxidase In Situ Apoptosis Detection Kit, The ApopTag Peroxidase In Situ Apoptosis Detection Kit detects apoptotic cells in situ by labeling & detecting DNA strand breaks by the TUNEL method.
Sigma-Aldrich
Anti-FAH, IgG fraction of antiserum
Roche
cOmplete, Mini Protease Inhibitor Cocktail, Tablets provided in a glass vial