Merck
  • Rapid differentiation of the human RPE cell line, ARPE-19, induced by nicotinamide.
Rapid differentiation of the human RPE cell line, ARPE-19, induced by nicotinamide.
Experimental eye research (2018-10-20)
Roni A Hazim, Stefanie Volland, Alice Yen, Barry L Burgess, David S Williams
ABSTRACT
Human RPE cell lines, especially the ARPE-19 cell line, are widely-used in eye research, as well as general epithelial cell studies. In comparison with primary RPE cells, they offer relative convenience and consistency, but cultures derived from these lines are typically not well differentiated. We describe a simple, rapid method to establish cultures from ARPE-19 cells, with significantly improved epithelial cell morphology and cytoskeletal organization, and RPE-related functions. We identify the presence of nicotinamide, a member of the vitamin B family, as an essential factor in promoting the observed differentiation, indicating the importance of metabolism in RPE cell differentiation.
Sigma-Aldrich
Anti-Rhodopsin Antibody, clone 4D2, clone 4D2, from mouse
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Tubulin, Acetylated antibody produced in mouse, clone 6-11B-1, ascites fluid