Merck
  • [Streptozocin-induced Alzheimer's disease as an independent risk factor for the development of hyperglycemia in Wistar rats].
[Streptozocin-induced Alzheimer's disease as an independent risk factor for the development of hyperglycemia in Wistar rats].
Problemy endokrinologii (2020-03-24)
A V Stavrovskaya, D N Voronkov, E A Shestakova, A S Gushchina, A S Olshansky, N G Yamshikova
ABSTRACT
In recent years the theme of the relationship of Alzheimers disease (AD) and metabolic disorders has been widely discussed. Nevertheless, it remains unclear whether AD is a direct cause of carbohydrate metabolism disorders or it is the presence of classical risk factors for type 2 diabetes mellitus (DM 2), primarily obesity, that significantly increases the risk of AD. To evaluate the separate contribution of two factors to the development of disorders of carbohydrate metabolism: (1) weight gain due to a high-calorie diet and (2) experimental-induced AD. Male Wistar rats were injected with streptozocin (STZ) in the lateral ventricles of the brain to induce AD or saline (sham operated animals - SO) during stereotactic operations. After 2 weeks, the animals were divided into four groups: 1) the SO group, which was assigned to the normal calorie (NCD) diet (SO NCD); 2) the SO group, which was assigned to the high-calorie diet (SO HCD); 3) the group to which the norm-calorie diet was prescribed after the administration of STZ into the lateral ventricles of the brain (STZ NCD); 4) the group to which the HCD was assigned after the administration of STZ (STZ HCD). The animals were on a diet for 3 months. Intraperitoneal glucose tolerance tests were carried out before the diet and after 3 months. At the end of the study, a morphological assessment of brain tissue, pancreas, and liver was performed. 3 months after surgical interventions and the appointment of diets, the glycemic curves significantly differed in the 4 studied groups: normoglycemia persisted only in the SO + NCD group, while HCD and the STZ administration were accompanied by the development of hyperglycemia (p = 0.0001). The STZ + NСD group, which represented the isolated effect of AD, was also characterized by impaired carbohydrate metabolism. A morphological study showed that HCD leads to a more pronounced ectopic accumulation of fat in the liver and pancreas tissue than NCD. The administration of STZ, regardless of the diet, led to changes typical for the AD model an increase in the size of the ventricles of the brain, degeneration of white matter, and the accumulation of -amyloid in the hypothalamus. The STZ-induced brain damage typical for AD led to impaired carbohydrate metabolism regardless of diet and was an independent risk factor for hyperglycemia.
Sigma-Aldrich
Oil Red O, certified by the BSC, certified by the Biological Stain Commission
Sigma-Aldrich
Anti-Tyrosine Hydroxylase antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution, sufficient for 10 blots
Sigma-Aldrich
Monoclonal Anti-CNP antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL2871, purified immunoglobulin, buffered aqueous glycerol solution