Merck
  • Expression and regulation of tumor necrosis factor-α-induced protein-8-like 2 is associated with acute lung injury induced by myocardial ischemia reperfusion in diabetic rats.
Expression and regulation of tumor necrosis factor-α-induced protein-8-like 2 is associated with acute lung injury induced by myocardial ischemia reperfusion in diabetic rats.
Microvascular research (2020-04-26)
Qian Kong, Min Yuan, Tingqian Ming, Qing Fang, Xiaojing Wu, Xuemin Song
ABSTRACT
The purpose of the present study was to investigate the possible role of TIPE2 on acute lung injury (ALI) induced by myocardial ischemia/reperfusion (MIR) in diabetic rats. Sprague-Dawley (SD) rats were randomly separated into four groups: control+sham (C + sham); control+MIR (C + MIR); diabetes+sham (D + sham); diabetes+MIR (D + MIR). Diabetes was induced using streptozotocin. Eight weeks after diabetes induction, MIR was conducted. At 2 h after MIR, myocardial injury indices were assessed; arterial blood, bronchoalveolar lavage fluid (BALF) and lung tissues were collected for corresponding detection. Rats subjected to MIR showed serious ALI (estimated via pathological changes, lung injury score and Wet/Dry weight ratio), lung inflammation and pulmonary cell apoptosis compared with sham groups, especially in D + MIR group. Evaluation of protein expression in lung tissues showed that p-JNK and nuclear NF-κB p65 protein levels were higher in D + MIR group as compared with C + MIR group. Besides, either hyperglycemia or MIR can significantly upregulate TIPE2 protein levels. In conclusion, diabetic lungs are more susceptible to MIR. TIPE2 may involve in this pathological process, possibly through regulation of inflammation, oxidative stress and apoptosis.
Sigma-Aldrich
NuCLEAR Extraction Kit, For mammalian tissue or cultured cells
Sigma-Aldrich
Wright Stain solution, according to Wright