Merck
  • Alantolactone suppresses inflammation, apoptosis and oxidative stress in cigarette smoke-induced human bronchial epithelial cells through activation of Nrf2/HO-1 and inhibition of the NF-κB pathways.
Alantolactone suppresses inflammation, apoptosis and oxidative stress in cigarette smoke-induced human bronchial epithelial cells through activation of Nrf2/HO-1 and inhibition of the NF-κB pathways.
Respiratory research (2020-04-24)
Xiaomin Dang, Beibei He, Qian Ning, Ya Liu, Jianxin Guo, Gang Niu, Mingwei Chen
ABSTRACT
It is well established that airway remodeling and inflammation are characteristics for chronic obstructive pulmonary disease (COPD). Moreover, cigarette smoke extract (CSE) promots inflammation, apoptosis and oxidative stress in COPD. And, there is evidence suggested that alantolactone (ALT), a sesquiterpene lactone isolated from Inula helenium, plays an adverse role in inflammation, apoptosis and oxidative stress. However, few studies have investigated the function and mechanism of ALT treatment on the COPD pathological process. The levels of IL-1 β, TNF-α, IL-6 and IFN-γ were examined by ELISA. Cells' apoptosis and caspase-3 activity were detected by Cell Death Detection PLUS enzyme-linked immunosorbent assay and caspase-Glo 3/7 Assay, respectively. The content of malondialdehyde (MDA) and superoxide dismutase (SOD) were determined by using MDA and SOD assay kits. Reactive oxygen species (ROS) generation was measured by DCFH-DA assay. Protein expression was assayed by Western blot. In the present study, we aimed to observe the protective effects of ALT against inflammation, apoptosis and oxidative stress in human bronchial epithelial Beas-2B and NHBE cells. Our results showed that different doses of CSE exposure induced Beas-2B and NHBE cell inflammatory cytokines IL-1 β, TNF-α, IL-6 and IFN-γ expression, cell apoptosis, caspase-3 activity and mediated oxidative stress markers MDA, ROS and SOD levels, while ALT treatment counteracted the effects of CSE. Further studies suggested that ALT attenuated NF-κB pathway activation. ALT also activated the Nrf2/HO-1 signal pathway through promoting Nrf2 nuclear aggregation and downstream HO-1 protein expression. HO-1 inhibitor tin protoporphyrin IX (SnPP IX) reversed the effects of ALT on Beas-2B and NHBE cell inflammation, apoptosis and oxidative stress. The above results collectively suggested that ALT suppressed CSE-induced inflammation, apoptosis and oxidative stress by modulating the NF-ĸB and Nrf2/ HO-1 axis.
Sigma-Aldrich
Alantolactone, ≥98% (HPLC)
Alantolactone, phyproof® Reference Substance