Merck
  • Endophilin-A3 and Galectin-8 control the clathrin-independent endocytosis of CD166.
Endophilin-A3 and Galectin-8 control the clathrin-independent endocytosis of CD166.
Nature communications (2020-03-21)
Henri-François Renard, François Tyckaert, Cristina Lo Giudice, Thibault Hirsch, Cesar Augusto Valades-Cruz, Camille Lemaigre, Massiullah Shafaq-Zadah, Christian Wunder, Ruddy Wattiez, Ludger Johannes, Pierre van der Bruggen, David Alsteens, Pierre Morsomme
ABSTRACT
While several clathrin-independent endocytic processes have been described so far, their biological relevance often remains elusive, especially in pathophysiological contexts such as cancer. In this study, we find that the tumor marker CD166/ALCAM (Activated Leukocyte Cell Adhesion Molecule) is a clathrin-independent cargo. We show that endophilin-A3-but neither A1 nor A2 isoforms-functionally associates with CD166-containing early endocytic carriers and physically interacts with the cargo. Our data further demonstrates that the three endophilin-A isoforms control the uptake of distinct subsets of cargoes. In addition, we provide strong evidence that the construction of endocytic sites from which CD166 is taken up in an endophilin-A3-dependent manner is driven by extracellular galectin-8. Taken together, our data reveal the existence of a previously uncharacterized clathrin-independent endocytic modality, that modulates the abundance of CD166 at the cell surface, and regulates adhesive and migratory properties of cancer cells.
Sigma-Aldrich
Anti-SH3GL3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Transferrin-biotin labeled human, lyophilized powder containing sodium citrate
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
2-Deoxy-D-glucose, ≥98% (GC), crystalline
Sigma-Aldrich
Dobutamine hydrochloride, ≥98%
Sigma-Aldrich
N-Acetyl-D-lactosamine, ≥98% (TLC)
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2