Merck
Inhibition of matrix metalloproteinase-2 by PARP inhibitors.
Biochemical and biophysical research communications (2009-07-22)
Adrian C Nicolescu, Andrew Holt, Arulmozhi D Kandasamy, Pal Pacher, Richard Schulz
ABSTRACT
Matrix metalloproteinase-2 (MMP-2), a ubiquitously expressed zinc-dependent endopeptidase, and poly(ADP-ribosyl) polymerase (PARP), a nuclear enzyme regulating DNA repair, are activated by nitroxidative stress associated with various pathologies. As MMP-2 plays a detrimental role in heart injuries resulting from enhanced nitroxidative stress, where PARP and MMP inhibitors are beneficial, we hypothesized that PARP inhibitors may affect MMP-2 activity. Using substrate degradation assays to determine MMP-2 activity we found that four PARP inhibitors (3-AB, PJ-34, 5-AIQ, and EB-47) inhibited 64kDa MMP-2 in a concentration-dependent manner. The IC(50) values of PJ-34 and 5-AIQ were in the high micromolar range and comparable to those of known MMP-2 inhibitors doxycycline, minocycline or o-phenanthroline, whereas those for 3-AB and EB-47 were in the millimolar range. Co-incubation of PARP inhibitors with doxycycline showed an additive inhibition of MMP-2 that was significant for 3-AB alone. These data demonstrate that the protective effects of some PARP inhibitors may include inhibition of MMP-2 activity.
Sigma-Aldrich
5-AIQ hydrochloride, ≥97% (HPLC), solid
Sigma-Aldrich
EB-47, ≥98% (HPLC), solid
Sigma-Aldrich
PARP Inhibitor IX, EB-47, The PARP Inhibitor IX, EB-47 controls the biological activity of PARP. This small molecule/inhibitor is primarily used for Cell Structure applications.
Sigma-Aldrich
PJ-34 hydrochloride hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
3-Aminobenzamide, ≥99% (TLC)
Sigma-Aldrich
PARP Inhibitor VIII, PJ34, The PARP Inhibitor VIII, PJ34, also referenced under CAS 344458-15-7, controls the biological activity of PARP. This small molecule/inhibitor is primarily used for Cell Structure applications.
Sigma-Aldrich
PARP Inhibitor I, 3-ABA, The PARP Inhibitor I, 3-ABA, also referenced under CAS 3544-24-9, controls the biological activity of PARP. This small molecule/inhibitor is primarily used for Cell Structure applications.