Merck
  • Periprotein lipidomes of Saccharomyces cerevisiae provide a flexible environment for conformational changes of membrane proteins.
Periprotein lipidomes of Saccharomyces cerevisiae provide a flexible environment for conformational changes of membrane proteins.
eLife (2020-04-18)
Joury S van 't Klooster, Tan-Yun Cheng, Hendrik R Sikkema, Aike Jeucken, Branch Moody, Bert Poolman
ABSTRACT
Yeast tolerates a low pH and high solvent concentrations. The permeability of the plasma membrane (PM) for small molecules is low and lateral diffusion of proteins is slow. These findings suggest a high degree of lipid order, which raises the question of how membrane proteins function in such an environment. The yeast PM is segregated into the Micro-Compartment-of-Can1 (MCC) and Pma1 (MCP), which have different lipid compositions. We extracted proteins from these microdomains via stoichiometric capture of lipids and proteins in styrene-maleic-acid-lipid-particles (SMALPs). We purified SMALP-lipid-protein complexes by chromatography and quantitatively analyzed periprotein lipids located within the diameter defined by one SMALP. Phospholipid and sterol concentrations are similar for MCC and MCP, but sphingolipids are enriched in MCP. Ergosterol is depleted from this periprotein lipidome, whereas phosphatidylserine is enriched relative to the bulk of the plasma membrane. Direct detection of PM lipids in the 'periprotein space' supports the conclusion that proteins function in the presence of a locally disordered lipid state.
Product Description
Avanti
16:0-18:1 PA, powder
Avanti
16:0-18:1 PA, 1-palmitoyl-2-oleoyl-sn-glycero-3-phosphate (sodium salt), chloroform
Avanti
18:0-18:1 PG, 1-stearoyl-2-oleoyl-sn-glycero-3-phospho-(1′-rac-glycerol) (sodium salt), chloroform
Avanti
16:0-18:1 PE, Avanti Research - A Croda Brand
Avanti
Brain PS, Avanti Research - A Croda Brand
Avanti
18:0-18:1 PG, 1-stearoyl-2-oleoyl-sn-glycero-3-phospho-(1′-rac-glycerol) (sodium salt), powder
Avanti
18:1 Cardiolipin, Avanti Research - A Croda Brand
Avanti
18:1 Cardiolipin, Avanti Research - A Croda Brand
Avanti
Brain PS, Avanti Research - A Croda Brand