Merck
  • A single amino acid change in inhibitory killer cell Ig-like receptor results in constitutive receptor self-association and phosphorylation.
A single amino acid change in inhibitory killer cell Ig-like receptor results in constitutive receptor self-association and phosphorylation.
Journal of immunology (Baltimore, Md. : 1950) (2014-12-17)
Santosh Kumar, Pabak Sarkar, Malcolm J W Sim, Sumati Rajagopalan, Steven S Vogel, Eric O Long
ABSTRACT
Signaling by immunoreceptors is often initiated by phosphorylation of cytosolic tyrosines, which then recruit effector molecules. In the case of MHC class I-specific inhibitory receptors, phosphorylation of cytosolic tyrosine residues within ITIMs results in recruitment of a protein tyrosine phosphatase that blocks activation signals. Recent work showed that signaling by an HLA-C-specific killer cell Ig-like receptor (KIR) is independent of signaling by activation receptors. It is not known how ITIM phosphorylation is initiated and regulated. In this article, we show that substitution of His-36 in the first Ig domain of KIR2DL1 with alanine (KIR2DL1-H36A) resulted in constitutive KIR2DL1 self-association and phosphorylation, as well as recruitment of tyrosine phosphatase SHP-1. Furthermore, substitution of His-36 with a similar bulky amino acid, phenylalanine, maintained the receptor in its unphosphorylated state, suggesting that steric hindrance by the His-36 side chain prevents constitutive KIR2DL1 self-association and ITIM phosphorylation. The equally strong phosphorylation of KIR2DL1 and KIR2DL1-H36A after inhibition of tyrosine phosphatase by pervanadate suggested that KIR2DL1-H36A is selectively protected from dephosphorylation. We propose that KIR phosphorylation is controlled by the accessibility of ITIM to tyrosine phosphatases and that KIR binding to HLA-C must override the hindrance that His-36 puts on KIR2DL1 self-association. Expression of KIR2DL1-H36A on NK cells led to stronger inhibition of lysis of HLA-C(+) target cells than did expression of wild-type KIR2DL1. These results revealed that ITIM phosphorylation is controlled by self-association of KIR and that His-36 serves as a gatekeeper to prevent unregulated signaling through KIR2DL1.
Product Description
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial
Sigma-Aldrich
Anti-SHP-1/SHPTP-1 Antibody, Upstate®, from rabbit
Roche
Anti-GFP, from mouse IgG1κ (clones 7.1 and 13.1)