Merck
  • Life-long impairment of glucose homeostasis upon prenatal exposure to psychostimulants.
Life-long impairment of glucose homeostasis upon prenatal exposure to psychostimulants.
The EMBO journal (2019-11-22)
Solomiia Korchynska, Maria Krassnitzer, Katarzyna Malenczyk, Rashmi B Prasad, Evgenii O Tretiakov, Sabah Rehman, Valentina Cinquina, Victoria Gernedl, Matthias Farlik, Julian Petersen, Sophia Hannes, Julia Schachenhofer, Sonali N Reisinger, Alice Zambon, Olof Asplund, Isabella Artner, Erik Keimpema, Gert Lubec, Jan Mulder, Christoph Bock, Daniela D Pollak, Roman A Romanov, Christian Pifl, Leif Groop, Tomas Gm Hökfelt, Tibor Harkany
ABSTRACT
Maternal drug abuse during pregnancy is a rapidly escalating societal problem. Psychostimulants, including amphetamine, cocaine, and methamphetamine, are amongst the illicit drugs most commonly consumed by pregnant women. Neuropharmacology concepts posit that psychostimulants affect monoamine signaling in the nervous system by their affinities to neurotransmitter reuptake and vesicular transporters to heighten neurotransmitter availability extracellularly. Exacerbated dopamine signaling is particularly considered as a key determinant of psychostimulant action. Much less is known about possible adverse effects of these drugs on peripheral organs, and if in utero exposure induces lifelong pathologies. Here, we addressed this question by combining human RNA-seq data with cellular and mouse models of neuroendocrine development. We show that episodic maternal exposure to psychostimulants during pregnancy coincident with the intrauterine specification of pancreatic β cells permanently impairs their ability of insulin production, leading to glucose intolerance in adult female but not male offspring. We link psychostimulant action specifically to serotonin signaling and implicate the sex-specific epigenetic reprogramming of serotonin-related gene regulatory networks upstream from the transcription factor Pet1/Fev as determinants of reduced insulin production.
Sigma-Aldrich
Monoclonal Anti-Glucagon antibody produced in mouse, clone K79bB10, ascites fluid
Sigma-Aldrich
Anti-HTR1A antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-HTR1F antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Supelco
Serotonin, analytical standard