Merck
  • NPGPx-Mediated Adaptation to Oxidative Stress Protects Motor Neurons from Degeneration in Aging by Directly Modulating O-GlcNAcase.
NPGPx-Mediated Adaptation to Oxidative Stress Protects Motor Neurons from Degeneration in Aging by Directly Modulating O-GlcNAcase.
Cell reports (2019-11-21)
Yung-Lin Hsieh, Fang-Yi Su, Li-Kai Tsai, Chien-Chang Huang, Yi-Ling Ko, Li-Wen Su, Kai-Yun Chen, Hsiu-Ming Shih, Chun-Mei Hu, Wen-Hwa Lee
ABSTRACT
Amyotrophic lateral sclerosis (ALS), the most common motor neuron disease, usually occurs in middle-aged people. However, the molecular basis of age-related cumulative stress in ALS pathogenesis remains elusive. Here, we found that mice deficient in NPGPx (GPx7), an oxidative stress sensor, develop ALS-like phenotypes, including paralysis, muscle denervation, and motor neurons loss. Unlike normal spinal motor neurons that exhibit elevated O-GlcNAcylation against age-dependent oxidative stress, NPGPx-deficient spinal motor neurons fail to boost O-GlcNAcylation and exacerbate ROS accumulation, leading to cell death. Mechanistically, stress-activated NPGPx inhibits O-GlcNAcase (OGA) through disulfide bonding to fine-tune global O-GlcNAcylation. Pharmacological inhibition of OGA rescues spinal motor neuron loss in aged NPGPx-deficient mice. Furthermore, expression of NPGPx in ALS patients is significantly lower than in unaffected adults. These results suggest that NPGPx modulates O-GlcNAcylation by inhibiting OGA to cope with age-dependent oxidative stress and protect motor neurons from degeneration, providing a potential therapeutic axis for ALS.
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Roche
Cell Proliferation Kit II (XTT), liquid, pkg of 1 kit, suitable for cell analysis, suitable for tissue culture
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Sigma-Aldrich
4-Nitrophenyl N-acetyl-β-D-glucosaminide, ≥99% (TLC)
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Ammonium nitrate, suitable for plant cell culture