Merck
  • Ceramide Content in Liver Increases Along with Insulin Resistance in Obese Patients.
Ceramide Content in Liver Increases Along with Insulin Resistance in Obese Patients.
Journal of clinical medicine (2019-12-18)
Hady Razak Hady, Agnieszka U Błachnio-Zabielska, Łukasz Szczerbiński, Piotr Zabielski, Monika Imierska, Jacek Dadan, Adam J Krętowski
ABSTRACT
The liver plays a central role in the glucose and lipid metabolism. Studies performed on animal models have shown an important role of lipid accumulation in the induction of insulin resistance. We sought to explain whether in obese humans, the insulin resistance is associated with hepatic ceramide accumulation. The experiments were conducted on obese men and women. Each gender was divided into three groups: Normal glucose tolerance group (NGT), Impaired glucose tolerance group (IGT), and Type 2 diabetic subjects (T2D). Ceramide (Cer) content was analyzed with the use of LC/MS/MS. An oral glucose tolerance test (OGTT), glycosylated hemoglobin (HbA1c), percentage body fat (FAT%), and body mass index (BMI) was also measured. Total hepatic ceramide was significantly higher in T2D females as compared to NGT females (p < 0.05), whereas in males, total ceramide was significantly higher in IGT and T2D as compared to NGT (p < 0.05). In both, men and women, the highest increase in T2D subjects, was observed in C16:0-Cer, C18:0:-Cer, C22:0-Cer, and C24:0-Cer (p < 0.05) as compared to NGT group. Interestingly, glucose (at 0' and at 120' in OGTT) and HbA1c positively correlated with the ceramide species that most increased in T2D patients (C16:0-Cer, C18:0-Cer, C22:0-Cer, and C24:0-Cer). In men glucose and HbA1c significantly correlated with only C22:0-Cer. This is one of the few studies comparing hepatic ceramide content in severely obese patients. We found that, ceramide content increased in diabetic patients, both in men and women, and the content of ceramide correlated with glycemic parameters. These data indicate ceramide contribution to the induction of hepatic insulin resistance.
Product Description
Avanti
C24:1 Ceramide-d7 (d18:1-d7/24:1(15Z)), Avanti Research - A Croda Brand 860679P, powder
Avanti
C24 Ceramide-d7 (d18:1-d7/24:0), Avanti Research - A Croda Brand 860678P, powder
Avanti
C15 Ceramide-d7 (d18:1-d7/15:0), Avanti Research - A Croda Brand 860681P, powder
Avanti
C18 Ceramide-d7 (d18:1-d7/18:0), Avanti Research - A Croda Brand 860677P, powder
Avanti
C16 Ceramide-d7 (d18:1-d7/16:0), Avanti Research - A Croda Brand 860676P, powder
Avanti
C24:1 Ceramide-d7 (d18:1-d7/24:1), Avanti Research - A Croda Brand 860679L-1MG860746L-1MG
Avanti
C20 Ceramide (d17:1/20:0), Avanti Research - A Croda Brand 860648P, powder