Merck
  • Increased lymphocyte activation and atherosclerosis in CD47-deficient mice.
Increased lymphocyte activation and atherosclerosis in CD47-deficient mice.
Scientific reports (2019-07-25)
Daniel Engelbertsen, Anu Autio, Robin A F Verwilligen, Marie A C Depuydt, Gail Newton, Sara Rattik, Erik Levinsohn, Gurpanna Saggu, Petr Jarolim, Huan Wang, Francisco Velazquez, Andrew H Lichtman, Francis W Luscinskas
ABSTRACT
CD47, also known as integrin-associated protein (IAP), is a transmembrane protein with multiple biological functions including regulation of efferocytosis and leukocyte trafficking. In this study we investigated the effect of CD47-deficiency on atherosclerosis using a model of adeno-associated virus (AAV)-induced hypercholesterolemia. We observed increased plaque formation in CD47 null mice compared to wild-type controls. Loss of CD47 caused activation of dendritic cells, T cells and natural killer (NK) cells, indicating an important role for CD47 in regulating immunity. In particular, Cd47 deficiency increased the proportion of IFN-γ producing CD90+ NK cells. Treatment with depleting anti-NK1.1 monoclonal antibody (mAb), but not depleting anti-CD4/CD8 mAbs, equalized atherosclerotic burden, suggesting NK cells were involved in the enhanced disease in Cd47 deficient mice. Additional studies revealed that levels of CD90+ and IFN-γ+ NK cells were expanded in atherosclerotic aorta and that CD90+ NK cells produce more IFN-γ than CD90- NK cells. Finally, we demonstrate that anti-CD47 (MIAP410) causes splenomegaly and activation of DCs and T cells, without affecting NK cell activation. In summary, we demonstrate that loss of CD47 causes increased lymphocyte activation that results in increased atherosclerosis.