Merck
  • Short-term exposure to dexamethasone promotes autonomic imbalance to the heart before hypertension.
Short-term exposure to dexamethasone promotes autonomic imbalance to the heart before hypertension.
Journal of the American Society of Hypertension : JASH (2018-06-28)
Francine Duchatsch, Paula B Constantino, Naiara A Herrera, Mayara F Fabrício, Lidieli P Tardelli, Aline M Martuscelli, Thiago J Dionísio, Carlos F Santos, Sandra L Amaral
ABSTRACT
Hypertension is one of the chronic side effects of dexamethasone (DEX) treatment; however, almost nothing is known about its acute effects. Therefore, the aim of this study was to investigate the possible mechanisms involved in blood pressure control after acute or short-term DEX treatment in adult animals. Eighty Wistar rats were divided into four groups: C1 and C5, for rats treated with saline for 1 or 5 days, respectively; D1 and D5, for rats treated with DEX for 1 or 5 days, respectively (decadron, 1 mg/kg, i.p.). Heart rate was increased in DEX treatment, but arterial pressure and cardiac muscle mass were not altered. Only few and isolated changes on gene expression and protein level of renin-angiotensin system components were observed. Five days of DEX treatment, but not one day, determined an increase in sympathetic component of spectral analysis (+75.93%, P < .05) and a significant reduction of parasympathetic component (-18.02%, P < .05), which contributed to the autonomic imbalance to the heart (LF/HF, +863.69%). The results of this present study demonstrated, for the first time, that short-term exposure to DEX treatment impairs the autonomic balance to the heart before hypertension, which was independent of renin-angiotensin system.
Sigma-Aldrich
Anti-AT2 Receptor Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-AT1 Receptor Antibody, Chemicon®, from rabbit