Merck
  • A new approach to examining the extraction process of Zhishi and Zhiqiao considering the synergistic effect of complex mixtures by PAMPA.
A new approach to examining the extraction process of Zhishi and Zhiqiao considering the synergistic effect of complex mixtures by PAMPA.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2018-09-22)
Hui Li, Honglian Zeng, Dan He, Menglei Wang, Linlin Liu, Wei Liang, Yisong Shu, Siyu Zhao, Guangyu Sun, Cheng Lv, Cheng Xiao, Yuanyan Liu
ABSTRACT
Zhishi (ZS) and Zhiqiao (ZQ) are two important traditional Chinese medicines (TCMs) that exert various pharmacological functions due to their active ingredients. However, the oral absorption of these ingredients requires further study. At the early drug discovery stage, the high-throughput parallel artificial membrane permeability assay (PAMPA) is one of the most frequently used to predict transcellular passive absorption in in-vitro models. This study aims to establish a new approach to examine an optimal extraction process that can take into account not only the concentration of active ingredients but also the overall absorption properties of the mixtures extracted from TCMs. A high-performance liquid chromatography triple-quadrupole mass spectrometry (HPLC-QqQ-MS/MS) method was validated for the determination of the effective permeability value (Pe) applied to the above experimental medium. The PAMPA experiment showed that certain active ingredients such as diosmin, rhoifolin, eriocitrin, narirutin, naringin, hesperidin and neohesperidin were not detected in the permeability assay of mono-constituents but were well detected and achieved a better absorption in the permeability assay of the mixture, indicating that certain unknown ingredients may act as cosolvents to improve the solubility or permeability of other ingredients. Furthermore, solid phase extraction (SPE) as an enrichment and purification process enhances absorption. In the present study, a novel in vitro approach was developed to decipher the potential role of TCMs in global absorption, and the extraction process for complex TCMs was described and systematically optimized.
Product Description
Supelco
Warfarin, analytical standard
Sigma-Aldrich
Auraptene, ≥98% (HPLC)
Sigma-Aldrich
Narirutin, ≥98% (HPLC)
Sigma-Aldrich
naringin dihydrochalcone, AldrichCPR