Merck
  • Development of an immunoaffinity column for the highly sensitive analysis of bisphenol A in 14 kinds of foodstuffs using ultra-high-performance liquid chromatography tandem mass spectrometry.
Development of an immunoaffinity column for the highly sensitive analysis of bisphenol A in 14 kinds of foodstuffs using ultra-high-performance liquid chromatography tandem mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2018-02-25)
Kai Yao, Kai Wen, Wenchong Shan, Sanlei Xie, Tao Peng, Jianyi Wang, Haiyang Jiang, Bing Shao
ABSTRACT
An immunoaffinity clean-up material based on a monoclonal antibody (mAb) has been prepared for concentrating and purifying bisphenol A (BPA) in 14 kinds of foodstuffs at trace level. Haptens and immunogen of bisphenol A have been synthesized and comprehensively characterized. An mAb towards BPA was prepared and cross-reactivities with 14 BPA analogues were below 5%. The prepared antibody was coupled to N-hydroxysuccinimide-activated Sepharose 4B to manufacture an immunoaffinity column (IAC), which was applied to purify BPA in 14 kinds of foodstuffs. The analyte was then detected by means of ultra-high-performance liquid chromatography tandem mass spectrometry (UPLC-MS/MS). Under the optimized conditions, compared with two traditional SPE clean-up methods, the IAC showed better selectivity (matrix effect <16.8%) and higher sensitivity. The limits of detection for BPA in 14 kinds of foodstuffs ranged from 0.001 μg L-1 to 0.01 μg kg-1, and the limits of quantification were in the range from 0.003 μg L-1 to 0.04 μg kg-1. The recoveries of BPA from spiked samples ranged from 82.0% to 104.9%, with RSDs below 13.8%. Besides, the IAC exhibited good reusability, with 40% column capacity remaining and no significant loss of recovery after 25 application cycles in real sample detection. These results demonstrated that the developed IAC-UPLC-MS/MS approach has wide applicability for purifying and detecting BPA in various foodstuffs.
Sigma-Aldrich
Ethyl 4-bromobutyrate, 95%
Sigma-Aldrich
Methyl 4-(bromomethyl)benzoate, 98%
Supelco
Bisphenol F bis(3-chloro-2-hydroxypropyl) ether, qualitative standard, mixture of 3 isomers ortho-ortho, ortho-para, para-para
Sigma-Aldrich
D.E.R. 332, used as embedding medium