Merck
  • Detailed qualitative analysis of honeybush tea (Cyclopia spp.) volatiles by comprehensive two-dimensional gas chromatography coupled to time-of-flight mass spectrometry and relation with sensory data.
Detailed qualitative analysis of honeybush tea (Cyclopia spp.) volatiles by comprehensive two-dimensional gas chromatography coupled to time-of-flight mass spectrometry and relation with sensory data.
Journal of chromatography. A (2017-09-06)
Gaalebalwe Ntlhokwe, Magdalena Muller, Elizabeth Joubert, Andreas G J Tredoux, André de Villiers
ABSTRACT
The volatile composition of honeybush (Cyclopia) species was studied by comprehensive two-dimensional gas chromatography coupled to time-of-flight mass spectrometry (GC×GC-TOF-MS). Headspace-solid phase micro-extraction (HS-SPME) was used to extract the volatile compounds from tea infusions prepared from the three species C. genistoides, C. maculata and C. subternata. A total of 287 compounds were identified, 101 of which were confirmed using reference standards, while the remainder were tentatively identified using mass spectral and retention index (RI) data. The identification power of TOF-MS enabled the tentative identification of 147 compounds for the first time in honeybush tea. The majority of the compounds identified were common to all three Cyclopia species, although there were differences in their relative abundances, and some compounds were unique to each of the species. In C. genistoides, C. maculata and C. subternata 265, 257 and 238 compounds were identified, respectively. Noteworthy was the tentative identification of cinnamaldehyde in particular C. maculata samples, which points to the likely contribution of this compound to their distinct sensory profiles. This study emphasises the complexity of honeybush tea volatile composition and confirms the power of GC×GC combined with TOF-MS for the analysis of such complex samples.
Product Description
Supelco
SPME fiber assortment kit 2, for use with autosampler, needle size 23 ga
Supelco
SPME StableFlex fiber assortment kit, needle size 23 ga, StableFlex fiber, for use with autosampler
Supelco
SPME fiber assortment kit 1, for use with autosampler, needle size 23 ga
Supelco
SPME fiber assortment kit 4, for use with autosampler, needle size 23 ga
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, for use with autosampler, needle size 23 ga, metal alloy fiber
Supelco
SPME StableFlex fiber assortment kit, for use with manual holder
Supelco
SPME fiber assortment kit 1, for use with manual holder, needle size 24 ga
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 24 ga, for use with autosampler
Supelco
SPME fiber assortment kit 1, for use with autosampler, needle size 24 ga
Supelco
SPME fiber assortment kit 4, for use with manual holder, size 24 ga
Supelco
SPME fiber assortment kit 2, for use with autosampler, size 24 ga
Supelco
SPME fiber assortment kit 4, for use with autosampler, size 24 ga
Supelco
SPME fiber assortment kit 2, for use with manual holder, needle size 24 ga
Supelco
SPME StableFlex fiber assortment kit, needle size 24 ga, for use with autosampler
Supelco
SPME fiber assortment kit 5, needle size 23 ga, for use with autosampler
Supelco
SPME fiber assembly polyacrylate (PA), df 85 μm(PA, for use with autosampler, needle size 24 ga
Supelco
SPME Portable Field Sampler, coating PDMS/DVB
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, for use with manual holder, needle size 24 ga
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 24 ga, for use with manual holder, StableFlex fiber
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, for use with autosampler, needle size 23 ga
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 30 μm(PDMS, needle size 24 ga, for use with manual holder
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 24 ga, for use with autosampler
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 23 ga, for use with autosampler
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, for use with manual holder, needle size 23 ga
Supelco
SPME Sampling Stand, for use with 15 mL vials
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 30 μm(PDMS, needle size 24 ga, for use with autosampler
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, for use with autosampler, needle size 23 ga
Supelco
SPME Fiber Assembly 50/30µm DVB/CAR/PDMS, needle size 24 ga, StableFlex fiber, for use with manual holder
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 24 ga, StableFlex, for use with autosampler
Supelco
SPME Portable Field Sampler, coating CAR/PDMS