Vanillin
Synonym(s):
Vanillin, 4-Hydroxy-3-methoxybenzaldehyde, Vanillic aldehyde
Linear Formula:
4-(HO)C6H3-3-(OCH3)CHO
CAS Number:
Molecular Weight:
152.15
Products
Product Category
Brand
Sustainability
Boiling Point (°C)
Feature
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
API Family
Reference Material Type
Special Grade
Technique
Showing 1-11 of 11
Compare
Product Number
Description
Pricing