
Triruthenium dodecacarbonyl

Synonym(s):
Ruthenium carbonyl, tri-Ruthenium dodecacarbonyl
Linear Formula:
Ru3(CO)12
Products
Product Category
Formula Weight
Physical Form
Purity
Quality Segment
Reaction Type
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing