
Trientine hydrochloride

Synonym(s):
Triethylenetetramine dihydrochloride, N,N′-Bis(2-aminoethyl)-1,2-ethanediamine dihydrochloride
Linear Formula:
NH2CH2CH2(NHCH2CH2)2NH2 · 2HCl
CAS Number:
Molecular Weight:
219.16
Products
Formula Weight
Manufacturer
Format
Application
Markush Class
Markush Group
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing