
trans-Cinnamaldehyde

Synonym(s):
trans-3-Phenyl-2-propenal
Linear Formula:
C6H5CH=CHCHO
CAS Number:
Molecular Weight:
132.16
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing