Skip to Content
Merck
Sulfolane 99%

Sulfolane

Sulfolane 99%
Synonym(s):
Tetrahydrothiophene 1,1-dioxide, Tetramethylene sulfone
Empirical Formula (Hill Notation):
C4H8O2S
CAS Number:
Molecular Weight:
120.17
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing